Learn More
Molex Molex-Polymicro Capillary 100UMx164UM 10M Reel
Supplier:  Molex 1068150020/10M
Encompass_Preferred
TSP100170 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 100um ID x 164um OD, spool length of 10m.
Catalog No. 50-110-7683