Learn More
Molex Molex-Polymicro Capillary 180UMx360UM 10M Reel
Supplier:  Molex 1068150025/10M
Encompass_Preferred
TSP180350 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 180um ID x 360um OD, spool length of 10m.
Catalog No. 50-110-7687