Learn More
Molex Molex-Polymicro Capillary 30UMx363UM 50M Reel
Supplier:  Molex 1068150013/50M
Encompass_Preferred
TSP030375 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 30um ID x 363um OD, spool length of 50m.
Catalog No. 50-110-8617