Learn More
Molex Molex-Polymicro Capillary 40UMx105UM 50M Reel
Supplier:  Molex 1068150596/50M
Encompass_Preferred
TSP040105 Flexible Fused Silica Capillary Tubing with Polyimide Coating: 40um ID x 105um OD, spool length of 50m.
Catalog No. 50-110-8618