Learn More
ABclonal Technology CAPN3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1199520UL
Encompass_Preferred
Calpain, a heterodimer consisting of a large and a small subunit, is a major intracellular protease, although its function has not been well established. This gene encodes a muscle-specific member of the calpain large subunit family that specifically binds to titin. Mutations in this gene are associated with limb-girdle muscular dystrophies type 2A. Alternate promoters and alternative splicing result in multiple transcript variants encoding different isoforms and some variants are ubiquitously expressed.
Catalog No. 50-001-15789