Learn More
Selleck Chemical LLC Capreomycin Sulfate S4234-50mg
SDP
Supplier:  Selleck Chemical LLC S423450MG
Encompass_Preferred
Capreomycin Sulfate is a cyclic peptide antibiotic and thought to inhibit protein synthesis by binding to the 70S ribosomal unit
Catalog No. 50-310-9117