Learn More
Medchemexpress LLC Capreomycin (sulfate) | 1405-37-4 | 99.52% | 5 G
SDP
Supplier:  Medchemexpress LLC HY175665G
Encompass_Preferred
Capreomycin sulfate is a macrocyclic peptide antibiotic. It functions by inhibiting phenylalanine synthesis in mycobacterial ribosomal translation. It also exhibits anti-amyloidogenic and pro-fibrinolytic activities, which means it can reduce cytotoxicity caused by amyloid by preventing amyloid fibrillation. This compound is used in the study of multidrug-resistant tuberculosis, type 2 diabetes, Alzheimer's disease, and Parkinson's disease.
  • Inhibits phenylalanine synthesis in mycobacterial ribosomal translation.
  • Exhibits anti-amyloidogenic activities.
  • Exhibits pro-fibrinolytic activities.
  • Reduces amyloid-induced cytotoxicity.
  • Prevents amyloid fibrillation.
  • Used in the study of multidrug-resistant tuberculosis.
  • Used in the study of type 2 diabetes.
  • Used in the study of Alzheimer's disease.
  • Used in the study of Parkinson's disease.
Catalog No. 50-003-87391