Learn More
TARGETMOL CHEMICALS INC AZD8797 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T143845MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. AZD8797 (KAND567) is an orally available, selective and potent human CX3CR1-converting antagonist with inhibitory effects on CX3CR1 and CXCR2, and potentially protects against SARS-CoV-2-induced neuronal damage, preventing worsening of nociceptive sensitization and microglial activation in a migraine model of rats following seizures. Purity 99.68%
Catalog No. 50-288-0498