Learn More
Medchemexpress LLC ε-caprolactone-d6 | 1219802-08-0 | 99.3% | 120.18 g·mol⁻¹ | C6H4D6O2 | 10 MG

Supplier: Medchemexpress LLC HYW015891S10MG
ε-Caprolactone-d6 is the deuterium-labeled form of ε-caprolactone supplied for laboratory research and analytical applications as a stable isotope reagent. It is provided with quality documentation and is intended for research use only.
- Deuterium-labeled stable isotope reagent.
- High purity (99.3%).
- Molecular formula C6H4D6O2; molecular weight 120.18 g·mol⁻¹.
- Supplied as a colorless to light yellow liquid.
- Available in small research sizes suitable for analytical workflows.
- Documentation includes datasheet, safety data sheet, and certificate of analysis.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.