Learn More
Microsolv Technology Corporation Caps, Headspace, Crimp Top, with Natural Silicone Rubber / Natural PTFE (5mil), High Purity Septa, fitted in Aluminum, Silver Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. 100/PK.
Supplier:  Microsolv Technology Corporation 950251121S
Encompass_Preferred
High Purity Silicone Rubber / PTFE Septa (5mil thick) with Durometer of 50 Shore A, (hardness) and 3.0mm thick.Wide temperature range from -60⁰ to 200⁰C.
Catalog No. 50-227-0359