Learn More
Microsolv Technology Corporation Caps, Screw Top, with Blue Silicone Rubber / White PTFE Pre-Slit Septa, fitted in "knurled", polypropylene, Blue Caps with 6mm holes. For use with 9-425 thread, 12x32mm autosampler vials. 100/PK.
Supplier:  Microsolv Technology Corporation 9502S33CB
Encompass_Preferred
Larger, 6mm hole in the cap provides a larger target area for autosamplers that might have difficulty and may strike the cap edge of smaller hole caps, during injection.
Catalog No. 50-227-0287