Learn More
Microsolv Technology Corporation Caps, Snap Top, with Silicone Rubber / PTFE, ultra-pure Septa, fitted in smooth, polyethylene, Light Blue Caps. For use with 11mm, 12x32mm, Snap Top autosampler vials. 100/PK.
Supplier:  Microsolv Technology Corporation 9509C10CB
Encompass_Preferred
This specially designed septa provides easy penetration by autosampler needles with improved resealing compared to other Snap Caps. The ultra-pure Silicone Rubber and PTFE have no added color producing the highest purity septa on the market with virtually no contamination or extractables. Recommended for LCMS/MS.
Catalog No. 50-227-0304