Learn More
Microsolv Technology Corporation Caps, Snap Top, with Silicone Rubber / PTFE, ultra-pure, Pre-Slit (Star Shape) Septa, fitted in smooth, polyethylene, Light Blue Caps. For use with 11mm, 12x32mm, Snap Top autosampler vials. 100/PK.
Supplier:  Microsolv Technology Corporation 9509C31CB
Encompass_Preferred
This specially designed septa provides easy penetration by autosampler needles with improved resealing compared to other Snap Caps. The ultra-pure Silicone Rubber and PTFE have no added color producing the highest purity septa on the market with virtually no contamination or extractables. The Star Shaped (*) slit offers a much greater target area for an autosampler needle to penetrate the septa and offers minimal resistance. It may not reseal as well as other Pre-Slit Septa. Recommended for LCMS/MS.
Catalog No. 50-227-0305