Learn More
Microsolv Technology Corporation Caps, Screw Top, Solid, with White Silicone Rubber / Beige PTFE Liner, fitted in knurled polypropylene, White Caps. For use with 24-400mm threaded EPA Style vials. ND24 threads. 100/PK.
Supplier:  Microsolv Technology Corporation 9704010CS
Encompass_Preferred
EPA quality liner is 45 Shore A, (Hardness), is 3.2mm thick and can be used for EPA testing or for long term storage with any 30, 40 or 60 EPA Style vial that has 40-400 threads. Solid top cap does not have an opening and is designed for storage or transportation.
Catalog No. 50-227-0285