Learn More
Microsolv Technology Corporation Caps, Screw Top, Solid, with a Red-Orange Silicone Rubber / TEF liner, in "knurled", closed Black Caps. For use with 10-425mm thread,12x32mm autosampler vials. 100/PK.
Supplier:  Microsolv Technology Corporation 9503S09KR
Encompass_Preferred
Use with wide mouth autosampler vials. The Silicone Rubber / TEF liner provides a very good seal for longer term storage in autosampler vials.
Catalog No. 50-227-1270