Learn More
Microsolv Technology Corporation Caps, Screw Top, Solid, with a Silicone Rubber / Polypropylene liner, in "knurled", closed Black Caps. 100/PK
Supplier:  Microsolv Technology Corporation 9502S09PS
Encompass_Preferred
This item has a minimum order quantity of 20 packs per supplier requirements.
Polyfluoroalkyl Substances (PFAS) can be tested or stored in our autosampler vials using these caps. The silicone rubber (for optimal sealing), and polypropylene fitted liners (for inertness) are excellent for storage of your sample before or after chromatography. These caps do not contain PTFE. Use with AQ™ Brand or RSA™ brand 9mm screw thread autosampler vials for best performance and sealing.
Catalog No. 50-226-9940