Learn More
Microsolv Technology Corporation Caps, Snap Top, with 0.04" thick, White Silicone Rubber / Natural PTFE Pre-Slit Septa, fitted in smooth, polyethylene, Light Blue Caps. For use with 11mm, 12x32mm Snap Top autosampler vials. 100/PK.
Supplier:  Microsolv Technology Corporation 9502CE35B
Encompass_Preferred
Pre-Slit Septa are normally recommended for Waters Alliance and Acquity instruments.Natural Septa does not have any color pigment and will not introduce extractables.
Catalog No. 50-227-0489