Learn More
Medchemexpress LLC Capsaicin | 404-86-4 | MFCD00017259 | 305.41 g·mol^-1 | C18H27NO3 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY10448200MG
Encompass_Preferred
Capsaicin is a naturally occurring capsaicinoid and a potent TRPV1 agonist derived from chili peppers. Supplied for research and analytical applications, it is commonly used to model nociception and to study pain, inflammation, and related signaling pathways. CAS 404-86-4; formula C18H27NO3; molar mass 305.41 g·mol-1.
  • Activates TRPV1 for receptor pharmacology studies.
  • Useful in pain, inflammation, and nociception models.
  • Applicable to in vitro biochemical and cell-based assays.
  • Provided as a small pack suitable for analytical and research use.
  • Well characterized with established chemical identifiers for traceability.
Catalog No. 50-002-42202