Learn More
Medchemexpress LLC Capsaicin β-D-glucopyranoside | 153409-16-6 | C24H37NO8 | 1 MG

Supplier: Medchemexpress LLC HY1534091MG
Capsaicin β-D-glucopyranoside is a glucoside converted by Capsaicin, the active ingredient of chili peppers responsible for their pungent flavor. This product is suitable for various research applications.
- Molecular weight: 467.55
- Appearance: Solid
- Color: White to off-white
- In vitro solubility: 100 mg/mL in DMSO (requires ultrasonication)
- Storage at -20°C, sealed, away from moisture and light
- In solvent storage at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.