Learn More
Cayman Chemical CapsaIcIn-d3 500ug
SDP
Supplier:  Cayman Chemical 34695500 UG
Encompass_Preferred
An internal standard for the quantification of capsaicin by GC- or LC-MS
Catalog No. 50-291-2363