Learn More
Indofine Chemical Capsaicin, Pure, 95%, 10 Mg
SDP
Supplier:  Indofine Chemical 028986 10 MG
Encompass_Preferred
CAPSAICIN, PURE 10 MG, Rare Organics & Biochemicals, Methyl-N-vanillyl-6-noneneamide, C18H27NO3, MW: 305.42, >95% by HPLC, 404-86-4, MFCD00017259
Catalog No. 50-908-897