Learn More
Selleck Chemical LLC Capsazepine-10mg
SDP
Supplier:  Selleck Chemical LLC S813710MG
Encompass_Preferred
Capsazepine acts as a competitive antagonist of capsaicin and a transient receptor potential vanilloid type 1(TRPV1) antagonist. If you want to do cell experiments, please select batch 01.
Catalog No. 50-239-0025