Learn More
Cayman Chemical Capsazepine Other Receptor Mod
SDP
Supplier:  Cayman Chemical 1000751810 MG
Encompass_Preferred
A competitive antagonist of TRPV1 which blocks the capsaicin-induced uptake of Ca2+ in neonatal rat dorsal root ganglia (IC50 = 0.42 µM) and Chinese hamster ovary cells (IC50 = 17 nM); does not block acid- or heat-induced activation of TRPV1 and may block receptors other than TRPV1
Catalog No. 50-289-0816