Learn More
Cayman Chemical CapsIconIate 10mg
SDP
Supplier:  Cayman Chemical 1817210 MG
Encompass_Preferred
A coniferyl ester that acts as a partial agonist of TRPV1 (EC50 = 3.2 µM), eliciting only 20% of the maximum response induced by capsaicin
Catalog No. 50-297-9805