Learn More
Braintree Scientific Capsule Sinker, for size 9el capsules
SDP
Supplier:  Braintree Scientific XSINK9EL
Encompass
MultipleCapsule Filler Fill 40 capsules at a time to withinin +/- 7-10% of average weight Includes filler, tamper, and spreader **Sold for research purposes only and not for human use**
Catalog No. 50-195-6156