Learn More
Medchemexpress LLC Captan | 133-06-2 | 99.65% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB158450MG
Encompass_Preferred
Captan is a common agricultural fungicide used to control Botrytis, Fusarium, Fusicoccum, Pythium. Captan enhances denitrifying and total culturable bacteria.
  • Used to control Botrytis.
  • Used to control Fusarium.
  • Used to control Fusicoccum.
  • Used to control Pythium.
  • Enhances denitrifying bacteria.
  • Enhances total culturable bacteria.
Catalog No. 50-004-11129