Learn More
CYDEX PHARMACEUTICALS A LIGAND COMPANY Captisol Research Grade, 5 kg
Supplier:  CYDEX PHARMACEUTICALS A LIGAND COMPANY RC0C7K05
Encompass
Captisol is a patent protected, uniquely modified cyclodextrin, whose chemical structure was rationally designed to enable the creation of new products by significantly improving solubility, stability, bioavailability and dosing of active pharmaceutical ingredients (APIs).
Catalog No. NC1457980
Only null left
Add to Cart
Request a Quote