Learn More
Selleck Chemical LLC Captopril-50mg
SDP
Supplier:  Selleck Chemical LLC S205150MG
Encompass_Preferred
Captopril (SQ 14225,Capoten) is an angiotensin-converting enzyme (ACE) inhibitor with IC50 of 6 nM.
Catalog No. 50-238-9589