Learn More
Medchemexpress LLC Captopril (Standard) | 62571-86-2 | 99.75% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB0368R
Encompass_Preferred
Captopril (Standard) is the analytical standard of Captopril, intended for research and analytical applications. It is an antihypertensive agent, a thiol-containing competitive, orally active angiotensin-converting enzyme (ACE) inhibitor (IC50=0.025 μM), and has been widely used for research of hypertension and congestive heart failure. It also acts as a New Delhi metallo-β-lactamase-1 (NDM-1) inhibitor with an IC50 of 7.9 μM.
  • Used as an analytical standard
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Angiotensin-converting enzyme (ACE) inhibitor
  • Used in hypertension and congestive heart failure research
  • New Delhi metallo-β-lactamase-1 (NDM-1) inhibitor
Catalog No. 50-004-07049