Learn More
Medchemexpress LLC Captopril (Standard) | 62571-86-2 | 99.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0368R100MG
Encompass_Preferred
Captopril (Standard) is the analytical standard of Captopril. This product is intended for research and analytical applications. Captopril (SQ 14225), an antihypertensive agent, is a thiol-containing competitive, orally active angiotensin-converting enzyme (ACE) inhibitor (IC50=0.025 μM) and has been widely used for research of hypertension and congestive heart failure. Captopril is also a New Delhi metallo-β-lactamase-1 (NDM-1) inhibitor with an IC50 of 7.9 μM.
  • Intended for research and analytical applications.
  • Thiol-containing competitive, orally active angiotensin-converting enzyme (ACE) inhibitor (IC50=0.025 μM).
  • Used for research of hypertension and congestive heart failure.
  • New Delhi metallo-β-lactamase-1 (NDM-1) inhibitor with an IC50 of 7.9 μM.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-07118