Learn More
Medchemexpress LLC HY-110404 5mg Medchemexpress, Capzimin CAS:2084868-04-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1104045MG
Encompass_Preferred
Medchemexpress, HY-110404 5mg Capzimin CAS:2084868-04-0 Capzimin is a potent and moderately specific proteasome isopeptidase Rpn11 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8797