Learn More
Medchemexpress LLC HY-119974 5mg Medchemexpress, Caracemide CAS:81424-67-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1199745MG
Encompass_Preferred
Medchemexpress, HY-119974 5mg Caracemide CAS:81424-67-1 Caracemide (NSC-253272) inhibits the enzyme ribonucleotide reductase of Escherichia coli. Caracemide is a novel anticancer agent derived from a hydroxamic acid and has demonstrated to produce severe central nervous system (CNS) toxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9563