Learn More
Medchemexpress LLC Carazolol-d7 | 1173021-02-7 | MFCD16652536 | 305.42 g/mol | C18H15D7N2O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY107327S5MG
Encompass_Preferred
Carazolol-d7 is the deuterium-labeled analogue of carazolol supplied as a 5 mg analytical reference standard. It is intended for use as an isotope-labeled internal standard in quantitative LC-MS and GC-MS assays, and for analytical method development and validation in research laboratories.
  • Deuterium-labeled internal standard for LC-MS and GC-MS
  • Molecular formula C18H15D7N2O2; molecular weight 305.42 g/mol
  • Supplied as a 5 mg reference material
  • Suitable for method development and quantitative analysis
  • Characterized for identity to support analytical use
Catalog No. 50-002-40671