Learn More
Medchemexpress LLC Carazolol-d7 | 1173021-02-7 | MFCD16652536 | >98.0% | 305.42 | C18H15D7N2O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY107327S1MG
Encompass_Preferred
Carazolol-d7 is the deuterium-labeled analogue of carazolol provided as a high-purity stable isotope standard for research and analytical applications. It is intended for use as an internal standard in quantitative mass spectrometry and for method development in pharmacology, ADME, and bioanalytical studies.
  • Deuterium-labeled internal standard for LC-MS/MS and GC-MS assays.
  • High chemical purity suitable for quantitative analysis.
  • Characterized by molecular formula and molecular weight for accurate identification.
  • Provided as a small, convenient pack size for trace-level applications.
  • Suitable for pharmacokinetic and metabolism studies requiring isotopic standards.
Catalog No. 50-002-41521