Learn More
Cayman Chemical Carazolol-d7 25mg
SDP
Supplier:  Cayman Chemical 3556025 MG
Encompass_Preferred
An internal standard for the quantification of carazolol by GC- or LC-MS
Catalog No. 50-291-3572