Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carazolol hydrochloride | 51997-43-4 | MFCD03428052 | 334.84 | C18H23ClN2O2 | 50 MG
SDP

Supplier:  Medchemexpress LLC HYW51726450MG

Encompass_Preferred

Carazolol hydrochloride is the hydrochloride salt of carazolol, a potent β1/β2-adrenoceptor antagonist supplied for laboratory research. It is used in pharmacology and receptor signaling studies; handle according to the product SDS and local laboratory safety protocols.

  • Potent β1/β2-adrenoceptor antagonist.
  • Hydrochloride salt for improved aqueous solubility.
  • Suitable for receptor pharmacology and signaling studies.
  • Available in small package sizes for laboratory use.
  • Chemical formula: C18H23ClN2O2; molecular weight: 334.84 g/mol.
  • CAS number: 51997-43-4.

Catalog No. 50-002-84888


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.