Learn More
Waters Corp Waters Corporation Carbamate Analysis Column, 60 Å, 4 μm, 3.9 mm x 150 mm
Supplier:  Waters Corp WAT035577
Encompass_Preferred
This Carbamate Analysis Column guarantees analysis of carbamate pesticides, exceeding AOAC Method 985.23 sensitivity. It is ideal for reversed-phase separation of carbamate pesticides and related compounds.
  • Optimized for reversed-phase separation of carbamate pesticides.
  • Features durable, high-efficiency 4μm spherical silica-based stationary phase.
  • Enables analysis of N-methylcarbamate and N-methyl-carbamoyl oxine pesticides in environmental/food samples.
  • Addresses carbamate analysis challenges, including peak identity confirmation.
  • Accurately identifies carbamates in complex matrices, reducing false positives.
  • Ensures precise surface chemistry, reproducibility, and stability via proprietary bonding/packing with strict quality control.
  • Manufactured to ISO 9001 and cGMP standards.
Catalog No. 50-785-363