Learn More
Medchemexpress LLC Carbamazepine 10,11-epoxide | 36507-30-9 | 99.14% | 252.27 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW01337850MG
Encompass_Preferred
Carbamazepine 10,11-epoxide is an orally active metabolite of Carbamazepine. It has an anticonvulsant effect and can be used for the research of seizures.
  • Orally active metabolite
  • Has anticonvulsant effect
  • Suitable for research of seizures
Catalog No. 50-004-54398