Learn More
Medchemexpress LLC Carbamazepine 10,11 epoxide-d10 | 1219804-16-6 | 99.0% | 262.33 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW013378S11MG
Encompass_Preferred
Carbamazepine 10,11 epoxide-d10 is a deuterium labeled version of Carbamazepine 10,11 epoxide, intended for research use. It serves as a valuable tool in quantitative analysis for various scientific applications.
  • Functions as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-54325