Learn More
Medchemexpress LLC Carbamazepine-d10 | 132183-78-9 | 99.2% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0246S5MG
Encompass_Preferred
Carbamazepine-d10 is the deuterium-labeled Carbamazepine. Carbamazepine (CBZ), a sodium channel blocker, is an anticonvulsant agent. It is for research use only and not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06958