Learn More
Medchemexpress LLC Carbamic acid, N-[(1S)-1-[[[1-ethyl-3-[[3-(4-morpholinyl)propyl)amino]-2,3-dioxopropyl]amino]carbon | 160399-35-9 | 99.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1193781MG
Encompass_Preferred
AK 295 (CX 295) is a selective calpain inhibitor used in research to inhibit calpain-dependent apoptosis and provide neuroprotective effects. It is employed in studies of neurological, cardiovascular, and inflammatory diseases and is supplied with supporting documentation for laboratory use.
  • Selective calpain inhibitor for apoptosis and neuroprotection studies.
  • High purity suitable for biochemical and cellular assays.
  • Applicable to research on neurological and cardiovascular disease models.
  • Available in small milligram quantities for lab-scale experiments.
  • Certificate of analysis and safety data sheet available for quality assurance.
Catalog No. 50-002-34456