Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbamic acid, N-[2-[2-(2-aminoethoxy)ethoxy]ethyl]-bicyclo[6.1.0]non-4-yn-9-ylmethyl ester | 1263166-93-3 | MFCD19705418 | 97.0% | 324.42 g/mol | C17H28N2O4 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYW072752

Encompass_Preferred

endo-BCN-PEG2-NH2 is a PEG-based PROTAC linker containing a strained bicyclo[6.1.0]nonyne (BCN) alkyne and a terminal amine. It enables copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) for bioconjugation and is commonly used in PROTAC assembly and linker installation.

  • Contains BCN strained alkyne for SPAAC click chemistry.
  • Peg2 spacer with a terminal primary amine.
  • Purity: 97.0% (HPLC).
  • Physical form: colorless to light yellow oil.
  • Molecular weight: 324.42 g/mol; formula C17H28N2O4.
  • Storage: protect from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-08868


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.