Learn More
Medchemexpress LLC Carbamic acid, N-[2-[2-(2-bromoethoxy)ethoxy]ethyl]-, 1,1-dimethylethyl ester | 165963-71-3 | ≥98.0% | 312.20 | C11H22BrNO4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW006445500MG
Encompass_Preferred
N-Boc-PEG2-bromide is a Boc-protected PEG2-containing bromoalkyl reagent used as a spacer and functional handle in medicinal chemistry syntheses, commonly applied as a cleavable linker in antibody-drug conjugate construction and as a PROTAC linker where the terminal bromide enables nucleophilic substitution.
  • Provides a PEG2 spacer for improved solubility and flexibility.
  • Contains a terminal bromide for nucleophilic substitution reactions.
  • Boc-protected amine allows selective deprotection and further functionalization.
  • High purity (≥98.0%) suitable for research-grade synthesis.
  • Available in small-scale laboratory quantities such as 500 mg.
Catalog No. 50-002-02125