Learn More
Medchemexpress LLC Carbamic acid, N-[2-[2-(2-propyn-1-yloxy)ethoxy]ethyl]-, 1,1-dimethylethyl ester | 869310-84-9 | MFCD22418469 | 97.0% | 243.30 g/mol | C12H21NO4 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1188081G
Encompass_Preferred
Propargyl-PEG2-NHBoc is a PEG-based propargyl linker containing a Boc-protected amine and a terminal alkyne, designed for bioconjugation and linker synthesis in research applications. It is compatible with click chemistry and is supplied at research-scale quantities with high purity.
  • Contains terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Boc-protected primary amine for controlled deprotection and further functionalization.
  • PEG2 spacer improves aqueous solubility and flexibility for conjugation.
  • Suitable as a cleavable linker for ADC and PROTAC synthesis and general bioconjugation.
  • Supplied at high purity (≥97%) and available in small research quantities such as 1 G.
Catalog No. 50-002-28596