Learn More
Medchemexpress LLC Carbamic acid, N-[2-(2-aminoethoxy)ethyl]-, 1,1-dimethylethyl ester | 127828-22-2 | 204.27 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0082791G
Encompass_Preferred
Amino-PEG2-NH-Boc is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs, which are compounds designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. PROTACs consist of two ligands connected by a linker, one for an E3 ubiquitin ligase and another for the target protein.
  • Can be used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-47705