Learn More
Medchemexpress LLC Carbamic acid, N-[2-hydroxy-1-(hydroxymethyl)ethyl]-, 1,1-dimethylethyl ester | 125414-41-7 | MFCD00270213 | 97.0% | 191.23 g/mol | C8H17NO4 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01846410G
Encompass_Preferred
N-Boc-serinol is an alkyl/ether-based PROTAC linker intended for use in the synthesis of proteolysis-targeting chimeras (PROTACs). It is provided as a research-grade chemical with documented purity and batch-level analytical data to support synthetic and medicinal chemistry workflows.
  • Alkyl/ether-based linker structure suitable for PROTAC design.
  • Reported purity of 97.0%, supporting reliable synthetic use.
  • Well-characterized small molecule with known molecular formula and weight.
  • Available in multiple pack sizes to accommodate different experimental scales.
  • Supplied with datasheet and certificate of analysis for quality verification.
Catalog No. 50-002-99715