Learn More
Medchemexpress LLC Carbamic acid, N-[(2S,3R)-2-methyl-4-oxo-3-oxetanyl]-, [1,1'-biphenyl]-4-ylmethyl ester | 1439366-66-1 | 99.7% | 311.33 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY147773
Encompass_Preferred
NAAA-IN-1 (Compound 1) is a potent and selective inhibitor of NAAA with an IC50 of 7 nM. NAAA is a cysteine amidase which preferentially hydrolyzes the endogenous biolipids palmitoylethanolamide (PEA) and oleoylethanolamide (OEA). NAAA-IN-1 has the potential for the research of inflammation and pain.
  • Inhibitor type: potent and selective inhibitor of NAAA.
  • IC50: 7 nM.
  • Target: NAAA (N-acylethanolamine acid amidase), a cysteine amidase.
  • Function of NAAA: preferentially hydrolyzes endogenous biolipids palmitoylethanolamide (PEA) and oleoylethanolamide (OEA).
  • Research potential: potential for the research of inflammation and pain.
Catalog No. 50-003-62780