Learn More
Medchemexpress LLC Carbamic acid, N-(3-aminobicyclo[1.1.1]pent-1-yl)-, 1,1-dimethylethyl ester | 1638767-25-5 | MFCD27987307 | 100.0% | 198.27 g/mol | C10H18N2O2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW05442410G
Encompass_Preferred
tert-Butyl (3-aminobicyclo[1.1.1]pentan-1-yl)carbamate is a BOC-protected amine used as a synthetic intermediate in medicinal chemistry, providing a 3-aminobicyclo[1.1.1]pentane building block for incorporation into active compounds and linkers.
  • Used as a BOC-protected amine intermediate for drug synthesis.
  • High purity suitable for research use.
  • Molecular formula C10H18N2O2; molecular weight 198.27 g/mol.
  • Available in multiple pack sizes, including 10 g.
  • Suitable for incorporation into linkers and building blocks in medicinal chemistry.
Catalog No. 50-002-10913