Learn More
Medchemexpress LLC Carbamic acid, N-[6-(propylthio)-1H-benzimidazol-2-yl]-, methyl ester D3 | 1353867-92-1 | 98.3% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0223S5MG
Encompass_Preferred
Albendazole-d3 is a deuterium labeled form of Albendazole, a benzimidazole compound used to treat various worm infestations. It is intended for research use only.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Belongs to isotope-labeled compounds
  • Useful as an internal standard for clinical mass spectrometry, food, pesticides, and veterinary medicine
  • Isotope-labeled inhibitor for use in in vivo cell experiments
  • Relevant for therapeutic drug monitoring
  • Applied in disease areas such as infection and parasite studies
Catalog No. 50-004-06169