Learn More
Medchemexpress LLC Carbamic acid, N-(cis-3-hydroxycyclobutyl)-, phenylmethyl ester | 1403766-86-8 | MFCD23106304 | 99.6% | 221.26 g/mol | C12H15NO3 | 100 G

Supplier: Medchemexpress LLC HYW057811100G
Cis-benzyl 3-hydroxycyclobutylcarbamate is a carbamate-class chemical intermediate used in pharmaceutical research for the synthesis of various active compounds. The solid material is supplied with high reported purity and is available in multiple pack sizes including 100 G.
- Drug intermediate for synthesis of active compounds.
- High reported purity (about 99.6%).
- Molecular weight 221.26 g/mol.
- Molecular formula C12H15NO3.
- Appearance: white to off-white solid.
- Packaging available up to 100 G, larger quantities by inquiry.
- COA and SDS available for download.
- Recommended storage: 4°C sealed; in solvent storage conditions specified.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.